siRNA Details
Gene Symbol: Degs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053323.2 | NP_445775.2 | 2 | 532 | ||
XM_006250403.2 | XP_006250465.1 | 2 | 827 | |||
XM_006250404.2 | XP_006250466.1 | 2 | 773 | |||
GenBank mRNA | AJ938081.1 | - | 2 | 485 | ||
AY036902.1 | - | 2 | 514 | |||
BC083727.1 | - | 2 | 532 | 7191263 | ||
FQ225269.1 | - | 2 | 540 |
