siRNA Details
Gene Symbol: Rabggtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138708.2 | NP_619715.1 | 2 | 206 | ||
GenBank mRNA | BC060536.1 | - | 230 | |||
BC078683.1 | - | 2 | 195 | 7135106 | ||
FQ214773.1 | - | 210 | ||||
FQ216744.1 | - | 210 | ||||
FQ224712.1 | - | 209 | ||||
FQ229109.1 | - | 210 | ||||
L10416.1 | - | 2 | 206 | |||
S62097.1 | - | 2 | 206 |
