siRNA Details
Gene Symbol: Adprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183325.2 | NP_899154.2 | 4 | 924 | ||
XM_006248360.2 | XP_006248422.1 | 4 | 1140 | |||
GenBank mRNA | BC082065.1 | - | 4 | 924 | 7132643 | |
M86341.1 | - | 3 | 779 |
