siRNA Details
Gene Symbol: Asgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012503.2 | NP_036635.1 | 6 | 454 | ||
XM_006246579.1 | XP_006246641.1 | 6 | 566 | |||
XM_008767768.1 | XP_008765990.1 | 6 | 586 | |||
XM_008767769.1 | XP_008765991.1 | 6 | 530 | |||
GenBank mRNA | BC088154.1 | - | 6 | 454 | 7372227 | |
FQ209763.1 | - | 6 | 490 | |||
FQ218553.1 | - | 6 | 530 | |||
FQ218644.1 | - | 6 | 490 | |||
FQ218824.1 | - | 6 | 490 | |||
FQ219107.1 | - | 6 | 527 | |||
FQ219710.1 | - | 6 | 571 | |||
M21770.1 | - | 5 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
