siRNA Details
Gene Symbol: Atp2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012508.5 | NP_036640.1 | 4988 | |||
GenBank mRNA | J03754.1 | - | 4994 |

More Information
Other Species Targeted

Gene Ontology Categories:
