siRNA Details
Gene Symbol: Cacna1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012517.2 | NP_036649.2 | 1 | 280 | ||
XM_006237159.2 | XP_006237221.1 | 1 | 104 | |||
XM_006237170.2 | XP_006237232.1 | 1 | 125 | |||
XM_006237171.2 | XP_006237233.1 | 1 | 125 | |||
XM_006237172.2 | XP_006237234.1 | 1 | 125 | |||
GenBank mRNA | AF394938.1 | - | 1 | 131 | ||
AF394939.1 | - | 1 | 131 | |||
AF394940.1 | - | 1 | 131 | |||
M59786.1 | - | 1 | 283 |
