siRNA Details
Gene Symbol: Cacna1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012517.2 | NP_036649.2 | 21 | 3108 | ||
XM_006237159.2 | XP_006237221.1 | 22 | 3007 | |||
XM_006237171.2 | XP_006237233.1 | 21 | 2953 | |||
XM_006237175.2 | XP_006237237.1 | 22 | 3461 | |||
XM_008763216.1 | XP_008761438.1 | 22 | 4970 | |||
GenBank mRNA | AF394939.1 | - | 21 | 2959 | ||
AY974797.2 | - | 23 | 3225 | |||
DQ538522.1 | - | 23 | 2927 | |||
M59786.1 | - | 21 | 3111 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001255997, NM_001256001, NM_001255999, NM_001255998, NM_001256000, and others.
Gene Ontology Categories:
