siRNA Details
Gene Symbol: Gad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012563.1 | NP_036695.1 | 13 | 1429 | ||
GenBank mRNA | M72422.1 | - | 13 | 1429 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012563.1 | NP_036695.1 | 13 | 1429 | ||
GenBank mRNA | M72422.1 | - | 13 | 1429 |