siRNA Details
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012576.2 | NP_036708.2 | 8 | 2423 | ||
XM_008772065.1 | XP_008770287.1 | 8 | 2424 | |||
XM_008772066.1 | XP_008770288.1 | 9 | 2474 | |||
GenBank mRNA | AF455050.1 | - | 8 | 2415 | ||
AY066016.2 | - | 8 | 2418 | |||
M14053.1 | - | 8 | 2481 |
