siRNA Details
Gene Symbol: Nf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012609.1 | NP_036741.1 | 521 | |||
GenBank mRNA | D45201.1 | - | 521 |

More Information
Other Species Targeted

Gene Ontology Categories:
