siRNA Details
Gene Symbol: Nf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012609.1 | NP_036741.1 | 734 | |||
GenBank mRNA | D45201.1 | - | 734 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010897, L10370.
Gene Ontology Categories:
