siRNA Details
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239906.2 | 8, 9 | 1108 | |||
XM_006239907.2 | XP_006239969.1 | 8, 9 | 1098 | |||
XM_006239908.2 | XP_006239970.1 | 7, 8 | 1014 | |||
XM_008764573.1 | 8, 9 | 1081 | ||||
GenBank mRNA | BC078882.1 | - | 6, 7 | 786 | 7115603 | |
J04791.1 | - | 8, 9 | 902 | |||
M16982.1 | - | 8, 9 | 1166 |
