siRNA Details
Gene Symbol: Prnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012631.2 | NP_036763.1 | 3 | 475 | ||
XM_006235062.2 | XP_006235124.1 | 4 | 627 | |||
GenBank mRNA | BC072692.1 | - | 2 | 413 | 7099864 | |
FQ211780.1 | - | 555 | ||||
M20313.1 | - | 1 | 236 |
