siRNA Details
Gene Symbol: RT1-N1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012646.1 | NP_036778.1 | 5 | 916 |
Gene Symbol: RT1-N2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008854.2 | NP_001008854.1 | 5 | 916 | ||
XM_006255938.2 | XP_006256000.1 | 5 | 1230 |
Gene Symbol: RT1-N3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008855.2 | NP_001008855.1 | 5 | 1180 | ||
XM_006255908.2 | XP_006255970.1 | 5 | 2676 | |||
XM_006255909.2 | XP_006255971.1 | 5 | 2676 | |||
GenBank mRNA | BC158567.1 | - | 5 | 1153 | 9090368 | |
FQ228139.1 | - | 5 | 1072 | |||
L23128.1 | - | 5 | 1184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
