siRNA Details
Gene Symbol: Sult2a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012695.3 | NP_036827.3 | 5 | 750 | ||
GenBank mRNA | FQ209484.1 | - | 5 | 625 | ||
FQ209563.1 | - | 5 | 625 | |||
FQ210641.1 | - | 4 | 498 | |||
FQ218388.1 | - | 5 | 626 | |||
FQ218712.1 | - | 5 | 625 | |||
FQ219267.1 | - | 5 | 625 | |||
FQ219345.1 | - | 5 | 625 | |||
M33329.1 | - | 5 | 750 | |||
X63410.1 | - | 5 | 750 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
