siRNA Details
Gene Symbol: Itgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012711.1 | NP_036843.1 | 6 | 679 | ||
XM_006230214.2 | XP_006230276.1 | 7 | 799 | |||
GenBank mRNA | AF268593.1 | - | 6 | 679 | ||
U59801.1 | - | 2 | 164 |
