siRNA Details
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163168.2 | NP_001156640.1 | 2 | 800 | ||
NM_012731.2 | NP_036863.1 | 2 | 785 | |||
XM_006253507.2 | XP_006253569.1 | 2 | 736 | |||
XM_008771425.1 | XP_008769647.1 | 2 | 741 | |||
XM_008771426.1 | XP_008769648.1 | 2 | 627 | |||
XM_008771427.1 | XP_008769649.1 | 2 | 735 | |||
GenBank mRNA | AY265419.1 | - | 2 | 565 | ||
M55291.1 | - | 2 | 788 | |||
M55292.1 | - | 2 | 565 | |||
M55293.1 | - | 1 | 232 |
