siRNA Details
Gene Symbol: TTLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF104927.1 | - | 1540 | |||
AF173935.1 | - | 1653 | ||||
AL136687.1 | - | 1639 | ||||
BC014968.1 | - | 1626 | 4876418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
