siRNA Details
Gene Symbol: Cnr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012784.4 | NP_036916.1 | 2 | 1868 | ||
XM_006237984.2 | XP_006238046.1 | 2 | 2020 | |||
GenBank mRNA | X55812.1 | - | 2 | 1868 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
