siRNA Details
Gene Symbol: Dpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012789.1 | NP_036921.1 | 4 | 339 | ||
XM_008761879.1 | XP_008760101.1 | 5 | 848 | |||
GenBank mRNA | J02997.1 | - | 4 | 279 | ||
J04591.1 | - | 4 | 339 |
