siRNA Details
Gene Symbol: Fmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012792.1 | NP_036924.1 | 9 | 1848 | ||
XM_006250145.2 | XP_006250207.1 | 9 | 1935 | |||
XM_006250146.2 | XP_006250208.1 | 9 | 1832 | |||
GenBank mRNA | BC061567.1 | - | 9 | 1841 | 6887467 | |
FQ218919.1 | - | 9 | 2059 | |||
M84719.1 | - | 9 | 1848 |
