siRNA Details
Gene Symbol: Adcy6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270785.1 | NP_001257714.1 | 7 | 1451 | ||
GenBank mRNA | L01115.1 | - | 7 | 1594 | ||
M96160.1 | - | 7 | 1451 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adcy6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270785.1 | NP_001257714.1 | 7 | 1451 | ||
GenBank mRNA | L01115.1 | - | 7 | 1594 | ||
M96160.1 | - | 7 | 1451 |