siRNA Details
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017008.4 | NP_058704.1 | 5 | 485 | ||
GenBank mRNA | AB017801.1 | - | 5 | 451 | ||
BC059110.1 | - | 5 | 486 | 6919611 | ||
BC087743.1 | - | 5 | 454 | 7321006 | ||
DQ403053.1 | - | 3 | 341 | |||
M17701.1 | - | 4 | 440 | |||
X02231.1 | - | 5 | 481 |

More Information
Other Species Targeted

Gene Ontology Categories:
