siRNA Details
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017008.4 | NP_058704.1 | 7 | 917 | ||
GenBank mRNA | AB017801.1 | - | 7 | 883 | ||
BC059110.1 | - | 7 | 918 | 6919611 | ||
BC087743.1 | - | 7 | 886 | 7321006 | ||
M11561.1 | - | 1 | 44 | |||
M17701.1 | - | 6 | 872 | |||
M29341.1 | - | 2 | 140 | |||
X02231.1 | - | 7 | 913 |

More Information
Other Species Targeted

Gene Ontology Categories:
