siRNA Details
Gene Symbol: Gsta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017013.4 | NP_058709.2 | 3, 4 | 200 | ||
XM_006243421.2 | XP_006243483.1 | 3, 4 | 267 | |||
XM_006243422.2 | XP_006243484.1 | 3, 4 | 261 | |||
GenBank mRNA | BC078706.1 | - | 3, 4 | 192 | 7128611 | |
FJ179397.1 | - | 2, 3 | 137 | |||
K01931.1 | - | 3, 4 | 200 |
Gene Symbol: Gsta5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010921.1 | NP_001010921.1 | 1, 2 | 163 | ||
GenBank mRNA | BC098898.1 | - | 1, 2 | 186 | 7322988 | |
CO566028.1 | - | 184 | ||||
K00136.1 | - | 1, 2 | 176 | |||
M25891.1 | - | 1, 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
