siRNA Details
Gene Symbol: Ldha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017025.1 | NP_058721.1 | 5 | 641 | ||
XM_006229232.2 | XP_006229294.1 | 5 | 809 | |||
GenBank mRNA | BC060587.1 | - | 5 | 593 | 6919582 | |
BC084698.1 | - | 4 | 543 | 7122339 | ||
BC168737.1 | - | 5 | 640 | 6917937 | ||
FQ214854.1 | - | 5 | 641 | |||
X01964.1 | - | 5 | 641 |
