siRNA Details
Gene Symbol: Mbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025291.1 | NP_001020462.1 | 7 | 753 | ||
NM_001025292.1 | NP_001020463.1 | 6 | 675 | |||
NM_001025293.1 | NP_001020464.1 | 6 | 630 | |||
NM_001025294.1 | NP_001020465.1 | 5 | 642 | |||
NM_017026.2 | NP_058722.1 | 5 | 552 | |||
XM_006254996.2 | XP_006255058.1 | 7 | 1105 | |||
XM_006254997.2 | XP_006255059.1 | 10 | 1415 | |||
XM_008772082.1 | XP_008770304.1 | 10 | 1430 | |||
XM_008772083.1 | XP_008770305.1 | 9 | 970 | |||
GenBank mRNA | AF439750.1 | - | 5 | 565 | ||
BC094522.1 | - | 5 | 508 | 7316167 | ||
FQ212549.1 | - | 1 | 31 | |||
JX131663.1 | - | 7 | 753 | |||
K00512.1 | - | 5 | 499 | |||
M25889.1 | - | 5 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
