siRNA Details
Gene Symbol: Glul | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017073.3 | NP_058769.4 | 7 | 2182 | ||
GenBank mRNA | BC061559.1 | - | 7 | 2195 | 6918514 | |
BC072694.1 | - | 7 | 2182 | 7103110 | ||
M29579.1 | - | 7 | 2193 | |||
M91652.1 | - | 7 | 2191 | |||
X07921.1 | - | 7 | 2193 |

More Information
Other Species Targeted

Gene Ontology Categories:
