siRNA Details
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017075.2 | NP_058771.2 | 12 | 1365 | ||
GenBank mRNA | D00512.1 | - | 12 | 1297 | ||
D13921.1 | - | 12 | 1297 | |||
FQ217320.1 | - | 12 | 1303 | |||
FQ228108.1 | - | 12 | 1351 |
