siRNA Details
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017093.1 | NP_058789.1 | 6 | 739 | ||
XM_008759108.1 | XP_008757330.1 | 7 | 3138 | |||
XM_008759109.1 | XP_008757331.1 | 6 | 726 | |||
XM_008759110.1 | XP_008757332.1 | 6 | 3003 | |||
XM_008759111.1 | XP_008757333.1 | 6 | 849 | |||
GenBank mRNA | D30041.1 | - | 6 | 739 |
