siRNA Details
Gene Symbol: MSRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135670.2 | NP_001129142.1 | 5 | 954 | ||
NM_001135671.2 | NP_001129143.1 | 6 | 1176 | |||
NM_001199729.2 | NP_001186658.1 | 7 | 1251 | |||
NM_012331.4 | NP_036463.1 | 6 | 1074 | |||
XM_017013448.1 | XP_016868937.1 | 5 | 1014 | |||
XM_017013449.1 | XP_016868938.1 | 6 | 1880 | |||
XM_017013450.1 | XP_016868939.1 | 6 | 817 | |||
XM_017013454.1 | XP_016868943.1 | 5 | 3186 | |||
GenBank mRNA | AF086925.1 | - | 6 | 893 | ||
AF183420.1 | - | 6 | 879 | |||
AJ242973.1 | - | 6 | 1038 | |||
AK123132.1 | - | 1 | 1466 | |||
AK293488.1 | - | 7 | 1251 | |||
AY690665.1 | - | 6 | 843 | |||
AY958429.1 | - | 6 | 1074 | |||
AY958430.1 | - | 7 | 1247 | |||
AY958431.1 | - | 6 | 1172 | |||
BC054033.1 | - | 6 | 1023 | 6504055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
