siRNA Details
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CB711853.1 | - | 3 | 199 | ||
CF978038.1 | - | 3 | 212 | |||
M11597.1 | - | 2 | 105 | |||
M26137.1 | - | 2 | 112 | |||
V01228.1 | - | 3 | 168 | |||
V01229.1 | - | 2 | 112 | |||
V01231.1 | - | 2 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
