siRNA Details
Gene Symbol: Calca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017338.2 | NP_059034.1 | 4 | 637 | ||
XM_008759676.1 | XP_008757898.1 | 4 | 688 | |||
GenBank mRNA | M26137.1 | - | 3 | 542 | ||
V01228.1 | - | 4 | 598 | |||
V01229.1 | - | 3 | 542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
