siRNA Details
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024823.4 | NP_001019994.4 | 5 | 2659 | ||
NM_001159653.1 | NP_001153125.1 | 1328 | ||||
NM_019132.1 | NP_062005.1 | 588 | ||||
NM_021845.5 | NP_068617.6 | 5 | 2659 | |||
GenBank mRNA | AF105254.1 | - | 1186 | |||
AF107844.1 | - | 1146 | ||||
AF184151.1 | - | 178 | ||||
BC061967.1 | - | 550 | ||||
BC158589.1 | - | 443 | 9099119 | |||
BC168156.1 | - | 452 | ||||
DQ120475.1 | - | 437 | ||||
DQ120476.1 | - | 437 | ||||
FQ212511.1 | - | 415 | ||||
FQ214546.1 | - | 327 | ||||
FQ219804.1 | - | 18 | ||||
FQ226208.1 | - | 116 | ||||
M12673.1 | - | 652 | ||||
M17525.1 | - | 588 | ||||
X84047.1 | - | 5 | 1807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
