siRNA Details
Gene Symbol: Gnas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024823.4 | NP_001019994.4 | 3013 | |||
NM_001159653.1 | NP_001153125.1 | 1682 | ||||
NM_019132.1 | NP_062005.1 | 942 | ||||
NM_021845.5 | NP_068617.6 | 3013 | ||||
GenBank mRNA | AF105254.1 | - | 1540 | |||
AF107844.1 | - | 1500 | ||||
BC061967.1 | - | 904 | ||||
BC158589.1 | - | 797 | 9099119 | |||
BC168156.1 | - | 806 | ||||
DQ120475.1 | - | 791 | ||||
DQ120476.1 | - | 791 | ||||
FQ214546.1 | - | 681 | ||||
FQ219804.1 | - | 372 | ||||
M12673.1 | - | 1006 | ||||
M17525.1 | - | 942 | ||||
X84047.1 | - | 2161 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_201618, NM_001077507, NM_010309, NM_001077510, NM_022000, and others.
Gene Ontology Categories:
