siRNA Details
Gene Symbol: Cxcl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022177.3 | NP_071513.1 | 3 | 688 | ||
GenBank mRNA | AF209976.1 | - | 3 | 639 | ||
BC078737.1 | - | 3 | 687 | 7128141 | ||
CK475409.1 | - | 3 | 687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
