siRNA Details
Gene Symbol: Scg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022669.1 | NP_073160.1 | 1 | 593 | ||
XM_006245148.2 | XP_006245210.1 | 2 | 783 | |||
XM_008767195.1 | XP_008765417.1 | 2 | 917 | |||
GenBank mRNA | AY168445.1 | - | 1 | 251 | ||
BC061549.1 | - | 2 | 635 | 6919444 | ||
M93669.1 | - | 1 | 593 | |||
X13618.1 | - | 1 | 593 |
