siRNA Details
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022922.2 | NP_075211.2 | 3, 4 | 353 | ||
GenBank mRNA | BC061781.1 | - | 3, 4 | 328 | ||
BC126087.1 | - | 3, 4 | 353 | 8368624 | ||
FQ214607.1 | - | 3, 4 | 438 | |||
FQ214670.1 | - | 3, 4 | 361 | |||
FQ214880.1 | - | 3, 4 | 366 | |||
FQ215213.1 | - | 2, 3 | 268 | |||
FQ215529.1 | - | 3, 4 | 365 | |||
FQ215606.1 | - | 2, 3 | 241 | |||
FQ215810.1 | - | 3, 4 | 366 | |||
FQ216226.1 | - | 2, 3 | 291 | |||
FQ216307.1 | - | 3, 4 | 252 | |||
FQ216584.1 | - | 3, 4 | 365 | |||
FQ224880.1 | - | 3, 4 | 364 | |||
FQ224920.1 | - | 3, 4 | 368 | |||
FQ224970.1 | - | 3, 4 | 361 | |||
L36250.1 | - | 3, 4 | 353 |
Gene Symbol: LOC100911515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750654.3 | XP_003750702.2 | 3, 4 | 509 |
Gene Symbol: LOC500959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033072.1 | NP_001028244.1 | 1 | 350 | ||
GenBank mRNA | AY461585.1 | - | 1 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
