siRNA Details
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022922.2 | NP_075211.2 | 6 | 581 | ||
GenBank mRNA | BC061781.1 | - | 6 | 556 | ||
BC126087.1 | - | 6 | 581 | 8368624 | ||
FQ214607.1 | - | 6 | 666 | |||
FQ214670.1 | - | 6 | 589 | |||
FQ214880.1 | - | 6 | 594 | |||
FQ215213.1 | - | 5 | 496 | |||
FQ215529.1 | - | 6 | 593 | |||
FQ215606.1 | - | 5 | 469 | |||
FQ215810.1 | - | 6 | 594 | |||
FQ216226.1 | - | 5 | 519 | |||
FQ216307.1 | - | 6 | 480 | |||
FQ216584.1 | - | 6 | 593 | |||
FQ223237.1 | - | 6 | 595 | |||
FQ224880.1 | - | 6 | 592 | |||
FQ224920.1 | - | 6 | 596 | |||
FQ224970.1 | - | 6 | 589 | |||
L36250.1 | - | 6 | 581 |
Gene Symbol: LOC100911515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750654.3 | XP_003750702.2 | 6 | 737 |
Gene Symbol: LOC500959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033072.1 | NP_001028244.1 | 1 | 578 | ||
GenBank mRNA | AY461585.1 | - | 1 | 578 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009415, AK150735, AK010808, X53333, AK212744, and others.
Gene Ontology Categories:
