siRNA Details
Gene Symbol: Agtr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M74054.1 | - | 3 | 1396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
