siRNA Details
Gene Symbol: C4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002805.2 | NP_001002805.1 | 14, 15 | 1908 |
Gene Symbol: C4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031504.3 | NP_113692.2 | 1913 | |||
GenBank mRNA | AY149995.1 | - | 1871 |
Gene Symbol: LOC103689965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772774.1 | XP_008770996.1 | 14, 15 | 1942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
