siRNA Details
Gene Symbol: Gsta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031509.2 | NP_113697.1 | 4, 5 | 515 | ||
GenBank mRNA | AF111160.1 | - | 4, 5 | 515 | ||
AF127460.1 | - | 3, 4 | 285 | |||
BC059128.1 | - | 4, 5 | 331 | 6922234 | ||
BC088127.1 | - | 4, 5 | 315 | 7322480 | ||
FJ179398.1 | - | 3, 4 | 276 | |||
FQ209644.1 | - | 4, 5 | 331 | |||
FQ211002.1 | - | 4, 5 | 326 | |||
FQ211022.1 | - | 4, 5 | 327 | |||
FQ213952.1 | - | 4, 5 | 466 | |||
FQ218347.1 | - | 4, 5 | 327 | |||
FQ218448.1 | - | 4, 5 | 324 | |||
FQ218456.1 | - | 4, 5 | 327 | |||
FQ218508.1 | - | 4, 5 | 326 | |||
FQ219165.1 | - | 4, 5 | 327 | |||
FQ219196.1 | - | 4, 5 | 327 | |||
FQ219451.1 | - | 4, 5 | 335 | |||
FQ219713.1 | - | 4, 5 | 327 | |||
FQ226984.1 | - | 4, 5 | 329 | |||
FQ229239.1 | - | 4, 5 | 327 | |||
K01932.1 | - | 4, 5 | 320 | |||
S72505.1 | - | 4, 5 | 425 | |||
X78848.1 | - | 4, 5 | 425 |
Gene Symbol: Gsta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009920.2 | NP_001009920.1 | 2, 3 | 504 | ||
NM_001159739.2 | NP_001153211.1 | 3, 4 | 474 | |||
NM_001287025.1 | NP_001273954.1 | 2, 3 | 366 | |||
GenBank mRNA | BC100080.1 | - | 3, 4 | 331 | 7324340 | |
CO566642.1 | - | 3, 4 | 337 | |||
DY308874.1 | - | 3, 4 | 398 | |||
DY573045.1 | - | 1, 2 | 199 | |||
FQ210706.1 | - | 2, 3 | 333 | |||
FQ218526.1 | - | 3, 4 | 347 | |||
FQ218726.1 | - | 3, 4 | 341 | |||
FQ219557.1 | - | 3, 4 | 341 | |||
FQ219615.1 | - | 3, 4 | 474 | |||
FQ219704.1 | - | 3, 4 | 341 | |||
FQ231428.1 | - | 3, 4 | 345 | |||
S72506.1 | - | 2, 3 | 352 | |||
S82820.1 | - | 2, 3 | 504 | |||
X78847.1 | - | 2, 3 | 352 |
Gene Symbol: LOC102550391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244659.2 | XP_006244721.2 | 510 | |||
GenBank mRNA | FQ218982.1 | - | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
