siRNA Details
Gene Symbol: Met | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031517.2 | NP_113705.1 | 1 | 204 | ||
XM_006236129.2 | XP_006236191.1 | 3 | 620 | |||
GenBank mRNA | U65007.1 | - | 1 | 204 | ||
X96786.1 | - | 1 | 204 |

More Information
Other Species Targeted

Gene Ontology Categories:
