siRNA Details
Gene Symbol: SF3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012433.3 | NP_036565.2 | 16 | 2346 | ||
GenBank mRNA | AF054284.1 | - | 16 | 2252 | ||
AK308122.1 | - | 16 | 2298 | |||
BC039020.1 | - | 16 | 2269 | |||
BC056155.1 | - | 16 | 2296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
