siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270589.1 | NP_001257518.1 | 2 | 736 | ||
NM_001270590.1 | NP_001257519.1 | 2 | 421 | |||
NM_001270591.1 | NP_001257520.1 | 3 | 199 | |||
NM_001270592.1 | NP_001257521.1 | 3 | 196 | |||
NM_001270593.1 | NP_001257522.1 | 2 | 133 | |||
NM_001270594.1 | NP_001257523.1 | 2 | 130 | |||
NM_134390.3 | NP_599217.1 | 2 | 739 | |||
XM_006236455.2 | XP_006236517.1 | 2 | 122 | |||
GenBank mRNA | AF370882.1 | - | 1 | 92 | ||
BC059116.1 | - | 2 | 238 | 6920949 | ||
FM070640.1 | - | 2 | 120 | |||
FM086181.1 | - | 3 | 183 | |||
FN804490.1 | - | 3 | 182 | |||
FQ212987.1 | - | 2 | 128 | |||
FQ219149.1 | - | 3 | 183 | |||
FQ219532.1 | - | 3 | 183 | |||
FQ219576.1 | - | 2 | 127 | |||
FQ220000.1 | - | 2 | 312 | |||
FQ233370.1 | - | 3 | 193 | |||
FQ235102.1 | - | 2 | 145 |
