siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270589.1 | NP_001257518.1 | 5 | 1095 | ||
NM_001270590.1 | NP_001257519.1 | 5 | 780 | |||
NM_001270591.1 | NP_001257520.1 | 6 | 558 | |||
NM_001270592.1 | NP_001257521.1 | 6 | 555 | |||
NM_001270593.1 | NP_001257522.1 | 5 | 492 | |||
NM_001270594.1 | NP_001257523.1 | 5 | 489 | |||
NM_134390.3 | NP_599217.1 | 5 | 1098 | |||
XM_006236455.2 | XP_006236517.1 | 5 | 481 | |||
GenBank mRNA | AF370882.1 | - | 4 | 451 | ||
BC059116.1 | - | 5 | 597 | 6920949 | ||
FM070640.1 | - | 5 | 479 | |||
FM086181.1 | - | 6 | 542 | |||
FN804490.1 | - | 6 | 541 | |||
FQ209634.1 | - | 5 | 486 | |||
FQ212987.1 | - | 5 | 487 | |||
FQ219149.1 | - | 6 | 542 | |||
FQ219532.1 | - | 6 | 542 | |||
FQ219576.1 | - | 5 | 486 | |||
FQ220000.1 | - | 5 | 671 | |||
FQ233370.1 | - | 6 | 552 | |||
FQ235102.1 | - | 5 | 504 |
