siRNA Details
Gene Symbol: Cyp3a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147206.2 | NP_671739.2 | 8 | 863 | ||
GenBank mRNA | BC086985.1 | - | 8 | 863 | 7125835 | |
U60085.2 | - | 8 | 839 |

More Information
Other Species Targeted

Gene Ontology Categories:
