siRNA Details
Gene Symbol: Obp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147215.2 | NP_671748.2 | 639 | |||
GenBank mRNA | J00738.1 | - | 639 | |||
X14552.1 | - | 1346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
