siRNA Details
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172222.2 | NP_757376.2 | 4 | 629 | ||
GenBank mRNA | AY149994.1 | - | 4 | 539 | ||
BC070923.1 | - | 4 | 629 | 7096131 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172222.2 | NP_757376.2 | 4 | 629 | ||
GenBank mRNA | AY149994.1 | - | 4 | 539 | ||
BC070923.1 | - | 4 | 629 | 7096131 |