siRNA Details
Gene Symbol: Klrb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173292.2 | NP_775414.2 | 4 | 276 | ||
GenBank mRNA | AF541943.1 | - | 4 | 276 | ||
EF100682.1 | - | 4 | 276 | |||
EF100684.1 | - | 4 | 276 | |||
FJ158034.1 | - | 4 | 276 | |||
U56936.1 | - | 462 |
Gene Symbol: Klrb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010964.1 | NP_001010964.1 | 4 | 292 | ||
XM_006237414.2 | XP_006237476.1 | 4 | 352 | |||
GenBank mRNA | EF100677.1 | - | 4 | 276 | ||
EF100681.1 | - | 4 | 276 | |||
EF100683.1 | - | 4 | 276 | |||
FJ416340.1 | - | 4 | 297 | |||
FQ232553.1 | - | 3 | 155 | |||
M62891.1 | - | 4 | 292 |
Gene Symbol: RGD2301395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135687.1 | NP_001129159.1 | 4 | 276 | ||
GenBank mRNA | FJ158033.1 | - | 4 | 276 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
